Fisher Chemical Murexide, Indicator Tablets, 1000 Tablets

Representative image only.
Fisher Chemical Mfr # M2731000 – Item # EW-88076-28
Quality and consistency your lab can count on
  • Reagents and solutions to meet your lab and research needs

PRODUCT OPTIONS

$1,286.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Empirical FormulaC8H8N6O6
  • ClassTitrants
  • FormTablets
  • SynonymsAmmonium Purpurate Acid
  • Formula Weight284.19
  • PackagingGlass Bottle
  • DOT InformationNot regulated by DOT
  • Storage SymbolGray
  • CAS Number3051-09-0
  • Boiling Point C (° C)1500
  • Melting Point C (° C)770
  • DescriptionMurexide, Indicator Tablets, 1000 Tablets
Storage Code Gray

Key Features

  • Reagents and solutions to meet your lab and research needs

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat