This item is now discontinued. Please browse similar items

Fisher Chemical N53500 Nickel (II) Chloride Hexahydrate (Cryst./Technical) (500g)

Representative image only.
Fisher Chemical Mfr # N53500 – Item # EW-88076-78

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeTechnical
  • Empirical FormulaNiCl2•6H2O
  • ClassSolids
  • ChemistrySolutions
  • SynonymsNickelous Chloride
  • FormCrystalline
  • Weight (g)500
  • Formula Weight237.71
  • PackagingGlass bottle
  • DOT InformationTOXIC SOLID, INORGANIC, N.O.S. (NICKEL(II) CHLORIDE HEXAHYDRATE); Class 6.1, Packing Group III
  • Storage SymbolBlue
  • Hazard SymbolToxic
  • CAS Number7791-20-0
  • UN NumberUN3288
  • DescriptionNickel (II) Chloride Hexahydrate (Cryst./Technical) (500g)

More About this Item


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat