Fisher Chemical O209320 Cyclohexane (Reagent) (20 liter)

Representative image only.
Fisher Chemical Mfr # O209320 – Item # EW-88077-08
Quality and consistency your lab can count on
Often bought with…

SPECS & DESCRIPTION

Specifications & Description

  • ChemistrySolvents
  • Volume (mL)20000
  • Formula Weight84.16
  • CAS Number110-82-7
  • DescriptionCyclohexane (Reagent) (20 liter)
FlammableHarmfulStorage Code Red

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat