Fisher Chemical O21091 Cyclohexanone (Certified) (1 liter) CAS 108-94-1

Representative image only.
Fisher Chemical Mfr # O21091 – Item # EW-88006-64
Quality and consistency your lab can count on
  • Reagents and solutions to meet your lab and research needs
  • Assay (by GC) — ≥98%
  • Boiling Range — 2°C incl. 155°C
  • Acid Content (as Formic Acid) — ≤0.05%
  • Residue after Evaporation — ≤0.05%
Often bought with…

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeCertified
  • Empirical FormulaC6H10O
  • Linear FormulaCH2(CH2)4CO
  • ClassGeneral Solutions
  • ChemistrySolutions
  • SynonymsKetohexamethylene
  • FormLiquid
  • Volume (mL)1000
  • Formula Weight98.15
  • PackagingAmber Glass
  • DOT InformationFlammable Liquid; Class 3
  • Storage SymbolRed
  • Hazard SymbolHarmful
  • CAS Number108941
  • UN NumberUN1915
  • DescriptionCyclohexanone (Certified) (1 liter) CAS 108-94-1
HarmfulStorage Code Red

Key Features

  • Reagents and solutions to meet your lab and research needs
  • Assay (by GC) — ≥98%
  • Boiling Range — 2°C incl. 155°C
  • Acid Content (as Formic Acid) — ≤0.05%
  • Residue after Evaporation — ≤0.05%

More About this Item

The Acros Organics brand delivers consistent, high-quality organics and fine chemicals at great savings. It also focuses on the evolving trends in organic chemistry, supporting research, development and product activities through a range of off-the-shelf products and expertise in sourcing, manufacturing and technology partnerships. Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat