This item is now discontinued. Please browse similar items

Fisher Chemical O235825 N,N-Diethyl-p-phenylenediamine Oxalate, Reagent; 25 g

Representative image only.
Fisher Chemical Mfr # O235825 – Item # EW-88077-26
Reagents and solutions to meet your lab and research needs
  • Assay (by titration) - ≥85%

SPECS & DESCRIPTION

Specifications & Description

  • Weight (g)25
  • Formula Weight418.55
  • CAS Number62637-92-7
  • DescriptionN,N-Diethyl-p-phenylenediamine Oxalate (Reagent) (25g)
HarmfulStorage Code Gray

Key Features

  • Assay (by titration) - ≥85%

More About this Item

Quality and consistency your lab can count on.


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat