Fisher Chemical O36884 1-Methyl-2-pyrrolidinone (Laboratory) (4 liter)

Representative image only.
Fisher Chemical Mfr # O36884 – Item # EW-88077-90
  • Color (APHA) - ≤50

PRODUCT OPTIONS

$380.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeLaboratory
  • Empirical FormulaC5H9NO
  • ClassReagent Solvents
  • ChemistrySolutions
  • FormLiquid
  • Volume (mL)4000
  • Formula Weight99.13
  • PackagingGlass bottle
  • DOT InformationNot regulated by DOT
  • Storage SymbolGray
  • Hazard SymbolIrritant
  • CAS Number872504
  • Description1-Methyl-2-pyrrolidinone (Laboratory) (4 liter)

Key Features

  • Color (APHA) - ≤50

More About this Item


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat