Fisher Chemical Potassium Bromide, Infrared Grade, 25 g

Representative image only.
Cole-Parmer Mfr # P22725 – Item # EW-88019-76
Consistent, high quality organics and fine chemicals at great savings
  • Reagents and solutions to meet your lab and research needs
  • Loss on Drying — ≤0.005%
  • IR Scan — To pass test
Often bought with…

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeInfrared
  • Empirical FormulaKBr
  • ClassReagent Salts
  • ChemistryAnalytical Reagents
  • FormSolid
  • SynonymsHydrobromic acid potassium salt, Bromide salt of potassium
  • Weight (g)25
  • Formula Weight119
  • PackagingGlass Bottle
  • DOT InformationNot regulated by DOT
  • Storage SymbolGray
  • CAS Number7758-02-3
  • Boiling Point C (° C)1435
  • Melting Point C (° C)734
  • DescriptionPotassium Bromide, Infrared Grade, 25 g
Storage Code Gray

Key Features

  • Reagents and solutions to meet your lab and research needs
  • Loss on Drying — ≤0.005%
  • IR Scan — To pass test

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat