Fisher Chemical P279212 Potassium Permanganate (Crystalline/Certified ACS) (2.5kg)

Representative image only.
Fisher Chemical Mfr # P279212 – Item # EW-88020-82
  • Assay — ≥99.0%
  • Insoluble Matter — ≤0.2%
  • Chloride and Chlorate (as Cl) — ≤0.005%
  • Nitrogen Compounds (as N) — ≤0.005%
  • Sulfate — ≤0.02%
  • Mercury (as Hg) — ≤0.05ppm
Often bought with…

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeCertified ACS
  • ChemistryAnalytical Reagents;Solutions
  • Weight (g)2500
  • Formula Weight158.04
  • CAS Number7722-64-7
  • DescriptionPotassium Permanganate (Crystalline/Certified ACS) (2.5kg)
HarmfulOxidizing

Key Features

  • Assay — ≥99.0%
  • Insoluble Matter — ≤0.2%
  • Chloride and Chlorate (as Cl) — ≤0.005%
  • Nitrogen Compounds (as N) — ≤0.005%
  • Sulfate — ≤0.02%
  • Mercury (as Hg) — ≤0.05ppm

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat