Fisher Chemical P333250LB Potassium Chloride (Cryst./EP/BP/USP/FCC) (250 lb.)

Representative image only.
Cole-Parmer Mfr # P333250LB – Item # EW-88079-10
Quality and consistency your lab can count on
  • Reagents and solutions to meet your lab and research needs
Often bought with…

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeBP, EP, FCC or USP
  • Empirical FormulaKCl
  • ClassReagent Salts
  • ChemistrySolutions
  • FormCrystalline
  • Weight (g)113398
  • Formula Weight74.55
  • PackagingFiber Drum
  • DOT InformationNot regulated by DOT
  • Storage SymbolGray
  • Hazard SymbolIrritant
  • CAS Number7447-40-7
  • DescriptionPotassium Chloride (Cryst./EP/BP/USP/FCC) (250 lb.)
IrritantStorage Code Gray

Key Features

  • Reagents and solutions to meet your lab and research needs

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat