Fisher Chemical Potassium Chloride, BP/EP/FCC/JP/USP Grade, 2.5 kg

Representative image only.
Fisher Chemical Mfr # P335212 – Item # EW-88019-88
Quality and consistency your lab can count on
  • Reagents and solutions to meet your lab and research needs
  • Meets USP Chemical Specifications
  • Assay (dried basis) — 99.0-100.5%
  • Acidity or Alkalinity — To pass test
  • Iodide or Bromide — To pass test
  • Heavy Metals (as Pb) — ≤0.001%
  • Sodium — To pass test
  • Loss on Drying at 105°C — ≤1.0%
  • Magnesium and Calcium — To pass test
  • Organic Volatile Impurities — Meets requirements
  • Bacterial Endotoxins — Report
  • Meets FCC Chemical Specifications
  • Assay (dried basis) — ≥99.0%
  • Acidity or Alkalinity — To pass test
  • Heavy Metals (as Pb) — ≤5mg/kg
  • Iodide or/and Bromide — To pass test
  • Loss on Drying — ≤1.0%
  • Sodium — To pass test
  • Meets EP and BP Chemical Specifications
  • Assay — 99.0-100.5%
  • Appearance of Solution — Clear and colorless
  • Acidity or Alkalinity — To pass test
  • Bromides — ≤0.1%
  • Iodides — To pass test
  • Sulfates — ≤300ppm
  • Barium — To pass test
  • Heavy Metals — ≤10ppm
  • Iron — ≤20ppm
  • Magnesium and Alkaline Earth Metals — ≤200ppm
  • Loss on Drying (105°C) — ≤1.0%
  • Meets JP Chemical Specifications
  • Assay (dried basis) — ≥99%
  • Clarity & Color of Solution — Clear and colorless
  • Acidity and Alkalinity — To pass test
  • Bromide — To pass test
  • Iodide — To pass test
  • Heavy Metals — ≤5ppm
  • Calcium and Magnesium&dot

PRODUCT OPTIONS

$736.50USD / Each
Often bought with…

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeBP/EP/FCC/JP/USP
  • Empirical FormulaKCl
  • ClassReagent Salts
  • ChemistryAnalytical Reagents
  • FormSolid
  • SynonymsKCl
  • Weight (g)2500
  • Formula Weight74.55
  • PackagingPoly Bottle
  • DOT InformationNot regulated by DOT
  • Storage SymbolGray
  • Hazard SymbolIrritant
  • CAS Number7447-40-7
  • Boiling Point C (° C)1420
  • Melting Point C (° C)770
  • DescriptionPotassium Chloride, BP/EP/FCC/JP/USP Grade, 2.5 kg
Storage Code Gray

Key Features

  • Reagents and solutions to meet your lab and research needs
  • Meets USP Chemical Specifications
  • Assay (dried basis) — 99.0-100.5%
  • Acidity or Alkalinity — To pass test
  • Iodide or Bromide — To pass test
  • Heavy Metals (as Pb) — ≤0.001%
  • Sodium — To pass test
  • Loss on Drying at 105°C — ≤1.0%
  • Magnesium and Calcium — To pass test
  • Organic Volatile Impurities — Meets requirements
  • Bacterial Endotoxins — Report
  • Meets FCC Chemical Specifications
  • Assay (dried basis) — ≥99.0%
  • Acidity or Alkalinity — To pass test
  • Heavy Metals (as Pb) — ≤5mg/kg
  • Iodide or/and Bromide — To pass test
  • Loss on Drying — ≤1.0%
  • Sodium — To pass test
  • Meets EP and BP Chemical Specifications
  • Assay — 99.0-100.5%
  • Appearance of Solution — Clear and colorless
  • Acidity or Alkalinity — To pass test
  • Bromides — ≤0.1%
  • Iodides — To pass test
  • Sulfates — ≤300ppm
  • Barium — To pass test
  • Heavy Metals — ≤10ppm
  • Iron — ≤20ppm
  • Magnesium and Alkaline Earth Metals — ≤200ppm
  • Loss on Drying (105°C) — ≤1.0%
  • Meets JP Chemical Specifications
  • Assay (dried basis) — ≥99%
  • Clarity & Color of Solution — Clear and colorless
  • Acidity and Alkalinity — To pass test
  • Bromide — To pass test
  • Iodide — To pass test
  • Heavy Metals — ≤5ppm
  • Calcium and Magnesium&dot

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat