Fisher Chemical P399SK4 Pentane (HPLC) (4 liter)

Representative image only.
Cole-Parmer Mfr # P399SK4 – Item # EW-88018-46
Quality and consistency your lab can count on
  • Reagents and solutions to meet your lab and research needs
  • Assay — ≥99%
  • Color (APHA) — ≤5
  • Fluorescence Background (as Quinine Sulfate) — To pass test
  • Refractive Index at 25°C — 1.3540-1.3570
  • Residue after Evaporation — ≤5ppm
  • Water — ≤0.02%
  • Optical Absorbance at 190nm — ≤1.00
  • at 200nm — ≤0.30
  • at 210nm — ≤0.10
  • at 254nm — ≤0.014
  • Submicron Filtered.
Often bought with…

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeHPLC
  • Empirical FormulaC5H12
  • Linear FormulaCH3(CH2)3CH3
  • ClassReagent Solvents
  • ChemistrySolutions
  • FormLiquid
  • Volume (mL)4000
  • Formula Weight72.15
  • PackagingSafety Coated Amber Glass Bottle
  • DOT InformationPentanes; Class 3, Packing Group II
  • Storage SymbolRed
  • Hazard SymbolEnvironment, Flammable, Harmful
  • CAS Number109-66-0
  • UN NumberUN1265
  • DescriptionPentane (HPLC) (4 L)
FlammableHarmfulStorage Code Red

Key Features

  • Reagents and solutions to meet your lab and research needs
  • Assay — ≥99%
  • Color (APHA) — ≤5
  • Fluorescence Background (as Quinine Sulfate) — To pass test
  • Refractive Index at 25°C — 1.3540-1.3570
  • Residue after Evaporation — ≤5ppm
  • Water — ≤0.02%
  • Optical Absorbance at 190nm — ≤1.00
  • at 200nm — ≤0.30
  • at 210nm — ≤0.10
  • at 254nm — ≤0.014
  • Submicron Filtered.

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat