Fisher Chemical S235500 Sodium Dichromate Dihydrate (Crystalline/Certified ACS) (500g)

Representative image only.
Fisher Chemical Mfr # S235500 – Item # EW-88023-92
  • Assay — ≥99.5%-100.5%
  • Insoluble Matter and Ammonium Hydroxide Precipitate — ≤0.005%
  • Chloride — ≤0.005%
  • Sulfate — ≤0.01%
  • Aluminum — ≤0.002%

PRODUCT OPTIONS

$526.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeCertified ACS
  • Empirical FormulaNa2Cr2O7&•;2H2O
  • ClassReagent Salts
  • ChemistrySolutions
  • FormCrystalline
  • SynonymsSodium Bichromate, Chromic Acid Disodium Salt
  • Weight (g)500
  • Formula Weight298.00
  • PackagingPoly Bottle
  • DOT InformationOXIDIZING SOLID, TOXIC, N.O.S.; Class 5.1 (6.1), Packing Group II
  • Storage SymbolYellow
  • Hazard SymbolEnvironment, Highly Toxic
  • CAS Number7789-12-0
  • UN NumberUN3087
  • DescriptionSodium Dichromate Dihydrate (Crystalline/Certified ACS) (500g)

Key Features

  • Assay — ≥99.5%-100.5%
  • Insoluble Matter and Ammonium Hydroxide Precipitate — ≤0.005%
  • Chloride — ≤0.005%
  • Sulfate — ≤0.01%
  • Aluminum — ≤0.002%
  • Calcium — ≤0.003%

More About this Item


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat