Fisher Chemical Sodium Carbonate Anhydrous, Certified ACS Grade, 1 kg

Representative image only.
Fisher Chemical Mfr # S2631 – Item # EW-88023-32
Quality and consistency your lab can count on
  • Reagents and solutions to meet your lab and research needs
  • Assay (dried basis) — ≥99.5%
  • Insoluble Matter — ≤0.01%
  • Loss on Heating at 285°C — ≤1.0%
  • Chloride — ≤0.001%
  • Nitrogen Compounds (as N) — ≤0.001%
  • Phosphate — ≤0.001%
  • Silica — ≤0.005%
  • Sulfur Compounds (as SO4) — ≤0.003%
  • Ammonium Hydroxide Precipitate — ≤0.01%
  • Calcium — ≤0.03%
  • Heavy Metals (as Pb) — ≤5ppm
  • Iron — ≤5ppm
  • Potassium — ≤0.005%
  • Magnesium — ≤0.005%

PRODUCT OPTIONS

$382.50USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeCertified ACS
  • Empirical FormulaNa2CO3
  • ClassReagent Salts
  • ChemistryAnalytical Reagents
  • FormSolid
  • SynonymsCarbonic Acid Disodium Salt, Soda Ash
  • Weight (g)1000
  • Formula Weight105.99
  • PackagingPoly Bottle
  • DOT InformationNot regulated by DOT
  • Storage SymbolGray
  • Hazard SymbolCorrosive, Irritant, Toxic
  • CAS Number497-19-8
  • Boiling Point C (° C)1600
  • Melting Point C (° C)854
  • DescriptionSodium Carbonate Anhydrous, Certified ACS Grade, 1 kg
IrritantStorage Code Gray

Key Features

  • Reagents and solutions to meet your lab and research needs
  • Assay (dried basis) — ≥99.5%
  • Insoluble Matter — ≤0.01%
  • Loss on Heating at 285°C — ≤1.0%
  • Chloride — ≤0.001%
  • Nitrogen Compounds (as N) — ≤0.001%
  • Phosphate — ≤0.001%
  • Silica — ≤0.005%
  • Sulfur Compounds (as SO4) — ≤0.003%
  • Ammonium Hydroxide Precipitate — ≤0.01%
  • Calcium — ≤0.03%
  • Heavy Metals (as Pb) — ≤5ppm
  • Iron — ≤5ppm
  • Potassium — ≤0.005%
  • Magnesium — ≤0.005%

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat