Fisher Chemical Sodium Hydrosulfite, Laboratory Grade, 500 g

Representative image only.
Fisher Chemical Mfr # S310500 – Item # EW-88080-66
Quality and consistency your lab can count on
  • Reagents and solutions to meet your lab and research needs

$179.25USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeLaboratory
  • Empirical FormulaNa2S2O4
  • ClassReagent Salts
  • ChemistryAnalytical Reagents
  • FormSolid
  • SynonymsSodium Dithionite
  • Weight (g)500
  • Formula Weight174.10
  • PackagingGlass Bottle
  • DOT InformationSodium Dithionite; Class 4.2, Packing Group II
  • Storage SymbolRed
  • Hazard SymbolHarmful
  • CAS Number7775-14-6
  • UN NumberUN1384
  • Melting Point C (° C)300
  • DescriptionSodium Hydrosulfite, Laboratory Grade, 500 g
HarmfulStorage Code Red

Key Features

  • Reagents and solutions to meet your lab and research needs

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat