Fisher Chemical S3201 Sodium Hydroxide (Pellets/NF/FCC) (1kg) CAS 1310-73-2

Representative image only.
Fisher Chemical Mfr # S3201 – Item # EW-88080-85
Quality and consistency your lab can count on
  • Reagents and solutions to meet your lab and research needs

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeFCC or NF
  • Empirical FormulaNaOH
  • ClassReagent Acids and Bases
  • SynonymsLye, Caustic Soda
  • FormPellets
  • Weight (g)1000
  • Formula Weight40.00
  • PackagingPoly Bottle
  • DOT InformationDOT Class 8
  • Storage SymbolWhite
  • Hazard SymbolCorrosive
  • CAS Number1310-73-2
  • UN NumberUN1823
  • DescriptionSodium Hydroxide (Pellets/NF/FCC) (1kg) CAS 1310-73-2
CorrosiveStorage Code White

Key Features

  • Reagents and solutions to meet your lab and research needs

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat