Fisher Chemical S3383 Sodium Nitrite (Flakes/USP/FCC) (3kg)

Representative image only.
Fisher Chemical Mfr # S3383 – Item # EW-88025-06
  • Assay (dried basis) — 97.0-101.0%
  • Loss on Drying — ≤0.25%
  • Heavy Metals (as Pb) — ≤0.002%
  • Lead — ≤10ppm

SPECS & DESCRIPTION

Specifications & Description

  • ChemistrySolutions
  • Weight (g)3000
  • Formula Weight69.00
  • CAS Number7632-00-0
  • DescriptionSodium Nitrite (Flakes/USP/FCC) (3kg)
OxidizingToxic

Key Features

  • Assay (dried basis) — 97.0-101.0%
  • Loss on Drying — ≤0.25%
  • Heavy Metals (as Pb) — ≤0.002%
  • Lead — ≤10ppm

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat