Fisher Chemical S3743 Sodium Phosphate Dibasic Anhydrous (Granular or Powder/Certified ACS) (3kg)

Representative image only.
Fisher BioReagents Mfr # S3743 – Item # EW-88025-32
Quality and consistency your lab can count on
  • Reagents and solutions to meet your lab and research needs
  • Assay — ≥99.0%
  • Insoluble Matter — ≤0.010%
  • Loss on Drying at 105°C — ≤0.2%
  • pH of a 5% Solution at 25°C — 8.7 to 9.3
  • Chloride — ≤0.002%
  • Nitrogen Compounds (as N) — ≤0.002%
  • Sulfate — ≤0.005%
  • Heavy Metals (as Pb) — ≤0.001%
  • Iron — ≤0.001%

SPECS & DESCRIPTION

Specifications & Description

  • ChemistrySolutions;Analytical Reagents
  • Weight (g)3000
  • Formula Weight141.96
  • CAS Number7558-79-4
  • DescriptionSodium Phosphate Dibasic Anhydrous (Granular or Powder/Certified ACS) (3kg)
Storage Code Gray

Key Features

  • Reagents and solutions to meet your lab and research needs
  • Assay — ≥99.0%
  • Insoluble Matter — ≤0.010%
  • Loss on Drying at 105°C — ≤0.2%
  • pH of a 5% Solution at 25°C — 8.7 to 9.3
  • Chloride — ≤0.002%
  • Nitrogen Compounds (as N) — ≤0.002%
  • Sulfate — ≤0.005%
  • Heavy Metals (as Pb) — ≤0.001%
  • Iron — ≤0.001%

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat