Fisher Chemical S637212 Sodium Bicarbonate (Crystalline Powder/USP/FCC/EP/BP/JP) (2.5kg)

Representative image only.
Mfr # S637212 – Item # EW-88023-04
Quality and consistency your lab can count on
  • Reagents and solutions to meet your lab and research needs
  • Meets USP Chemical Specifications
  • Assay (dried basis) — 99.0-100.5%
  • Ammonium — To pass test
  • Arsenic — ≤2ppm
  • Chloride — ≤150ppm
  • Heavy Metals (as Pb) — ≤5ppm
  • Loss on Drying — ≤0.25%
  • Insoluble Substances — To pass test
  • Normal Carbonate — To pass test
  • Limit of Sulfur Compounds — ≤0.015%
  • Organic Volatile Impurities — Meets requirements
  • Bacterial Endotoxins — Report
  • Meets FCC Chemical Specifications
  • Assay (dried basis) — 99.0-100.5%
  • Ammonia — To pass test
  • Heavy Metals — ≤5mg/kg
  • Insoluble Substances — To pass test
  • Loss on Drying — ≤0.25%
  • Meets EP and BP Chemical Specifications
  • Assay — 99.0-101.0%
  • Appearance of Solution — Clear and colorless
  • Carbonates — To pass test
  • Chlorides — ≤150ppm
  • Sulfates — ≤150ppm
  • Ammonium — ≤20ppm
  • Arsenic — ≤2ppm Calcium — ≤100ppm
  • Heavy Metals — ≤10ppm
  • Iron — ≤20ppm
  • Bacterial Endotoxins — Report
  • Meets JP Chemical Specifications
  • Assay (dried basis) — ≥99.0%
  • pH — 7.9-8.4
  • Clarity and Color of Solution — Clear and colorless
  • Chloride — ≤0.040%
  • Carbonate — To pass test
  • Ammonium — To pass test
  • Heavy Metals — ≤5ppm
  • Arsenic — ≤2ppm

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeBP, EP, FCC, JP, or USP
  • ChemistrySolutions
  • Weight (g)2500
  • Formula Weight84.01
  • CAS Number144-55-8
  • DescriptionSodium Bicarbonate (Crystalline Powder/USP/FCC/EP/BP/JP) (2.5kg)
Storage Code Gray

Key Features

  • Reagents and solutions to meet your lab and research needs
  • Meets USP Chemical Specifications
  • Assay (dried basis) — 99.0-100.5%
  • Ammonium — To pass test
  • Arsenic — ≤2ppm
  • Chloride — ≤150ppm
  • Heavy Metals (as Pb) — ≤5ppm
  • Loss on Drying — ≤0.25%
  • Insoluble Substances — To pass test
  • Normal Carbonate — To pass test
  • Limit of Sulfur Compounds — ≤0.015%
  • Organic Volatile Impurities — Meets requirements
  • Bacterial Endotoxins — Report
  • Meets FCC Chemical Specifications
  • Assay (dried basis) — 99.0-100.5%
  • Ammonia — To pass test
  • Heavy Metals — ≤5mg/kg
  • Insoluble Substances — To pass test
  • Loss on Drying — ≤0.25%
  • Meets EP and BP Chemical Specifications
  • Assay — 99.0-101.0%
  • Appearance of Solution — Clear and colorless
  • Carbonates — To pass test
  • Chlorides — ≤150ppm
  • Sulfates — ≤150ppm
  • Ammonium — ≤20ppm
  • Arsenic — ≤2ppm Calcium — ≤100ppm
  • Heavy Metals — ≤10ppm
  • Iron — ≤20ppm
  • Bacterial Endotoxins — Report
  • Meets JP Chemical Specifications
  • Assay (dried basis) — ≥99.0%
  • pH — 7.9-8.4
  • Clarity and Color of Solution — Clear and colorless
  • Chloride — ≤0.040%
  • Carbonate — To pass test
  • Ammonium — To pass test
  • Heavy Metals — ≤5ppm
  • Arsenic — ≤2ppm

More About this Item

The Acros Organics brand delivers consistent, high-quality organics and fine chemicals at great savings. It also focuses on the evolving trends in organic chemistry, supporting research, development and product activities through a range of off-the-shelf products and expertise in sourcing, manufacturing and technology partnerships. Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat