Fisher Chemical S642212 Sodium Chloride (Granular/USP/FCC/EP/BP/JP) (2.5kg)

Representative image only.
Cole-Parmer Mfr # S642212 – Item # EW-88023-58
Quality and consistency your lab can count on
  • Reagents and solutions to meet your lab and research needs
  • Meets USP Chemical Specifications
  • Assay (dried basis) — 99.0-100.5%
  • Appearance of Solution — To pass test
  • Acidity or Alkalinity — To pass test
  • Arsenic — ≤1ppm
  • Barium — To pass test
  • Magnesium and Alkaline Earth Metals (as Ca) — ≤0.01%
  • Heavy Metals (as Pb) — ≤5ppm
  • Iodides — To pass test
  • Bromides — ≤0.01%
  • Iron — ≤2ppm
  • Loss on Drying at 105°C — To pass test
  • Ferrocyanide — To pass test
  • Sulfate — ≤0.020%
  • Aluminum — ≤0.2ppm
  • Nitrites — To pass test
  • Limit of Phosphates — ≤0.0025%
  • Meets FCC Chemical Specifications
  • Assay (after drying at 625°C) — 97.5-100.5%
  • Arsenic — ≤1mg/kg
  • Calcium and Magnesium — ≤2.0%
  • Heavy Metals (as Pb) — ≤2mg/kg
  • Loss on Drying at 110°C — ≤0.5%
  • Meets EP and BP Chemical Specifications
  • Assay (dried basis) — 99.0-100.5%
  • Appearance of Solution — To pass test
  • Acidity and Alkalinity — To pass test
  • Bromides — ≤50ppm
  • Ferrocyanides — To pass test
  • Iodides — To pass test
  • Nitrites — To pass test
  • Phosphates — ≤25ppm
  • Sulfates — ≤200ppm (EP); ≤250ppm (BP)
  • Arsenic — ≤1ppm
  • Barium — To pass test
  • Heavy Metals (as Pb) — ≤5ppm (EP); ≤10ppm (BP)
  • Iron — ≤2ppm (EP); ≤20ppm (BP)
  • Magnesium and A

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeBP, EP, FCC, JP, or USP
  • Empirical FormulaNaCl
  • ClassReagent Salts
  • ChemistrySolutions
  • SynonymsSalt
  • FormPowder
  • Weight (g)2500
  • Formula Weight58.44
  • PackagingPoly Bottle
  • DOT InformationNot regulated by DOT
  • Storage SymbolGray
  • CAS Number7647-14-5
  • DescriptionSodium Chloride (Granular/USP/FCC/EP/BP/JP) (2.5 kg)
Storage Code Gray

Key Features

  • Reagents and solutions to meet your lab and research needs
  • Meets USP Chemical Specifications
  • Assay (dried basis) — 99.0-100.5%
  • Appearance of Solution — To pass test
  • Acidity or Alkalinity — To pass test
  • Arsenic — ≤1ppm
  • Barium — To pass test
  • Magnesium and Alkaline Earth Metals (as Ca) — ≤0.01%
  • Heavy Metals (as Pb) — ≤5ppm
  • Iodides — To pass test
  • Bromides — ≤0.01%
  • Iron — ≤2ppm
  • Loss on Drying at 105°C — To pass test
  • Ferrocyanide — To pass test
  • Sulfate — ≤0.020%
  • Aluminum — ≤0.2ppm
  • Nitrites — To pass test
  • Limit of Phosphates — ≤0.0025%
  • Meets FCC Chemical Specifications
  • Assay (after drying at 625°C) — 97.5-100.5%
  • Arsenic — ≤1mg/kg
  • Calcium and Magnesium — ≤2.0%
  • Heavy Metals (as Pb) — ≤2mg/kg
  • Loss on Drying at 110°C — ≤0.5%
  • Meets EP and BP Chemical Specifications
  • Assay (dried basis) — 99.0-100.5%
  • Appearance of Solution — To pass test
  • Acidity and Alkalinity — To pass test
  • Bromides — ≤50ppm
  • Ferrocyanides — To pass test
  • Iodides — To pass test
  • Nitrites — To pass test
  • Phosphates — ≤25ppm
  • Sulfates — ≤200ppm (EP); ≤250ppm (BP)
  • Arsenic — ≤1ppm
  • Barium — To pass test
  • Heavy Metals (as Pb) — ≤5ppm (EP); ≤10ppm (BP)
  • Iron — ≤2ppm (EP); ≤20ppm (BP)
  • Magnesium and A

More About this Item

The Acros Organics brand delivers consistent, high-quality organics and fine chemicals at great savings. It also focuses on the evolving trends in organic chemistry, supporting research, development and product activities through a range of off-the-shelf products and expertise in sourcing, manufacturing and technology partnerships. Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat