Fisher Chemical S648500 Sodium Formate (Cryst./Certified ACS) (500g)

Representative image only.
Fisher Chemical Mfr # S648500 – Item # EW-88083-05
  • Assay (HCOONa) - ≥99.0%
  • Insoluble Matter - ≤0.005%
  • Chloride - ≤0.001%
  • Sulfate - ≤0.001%
  • Calcium - ≤0.005%
  • Heavy Metals (as Pb) - ≤5ppm
  • Iron - ≤5ppm

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeACS Reagent
  • Empirical FormulaCHO2Na
  • Linear FormulaHCOONa
  • ClassSolids
  • ChemistryAnalytical Reagents;Solutions
  • FormCrystalline
  • SynonymsFormic acid sodium salt
  • Weight (g)500
  • Formula Weight68.01
  • PackagingAmber Glass
  • DOT InformationNot regulated by DOT
  • Storage SymbolGray
  • CAS Number141-53-7
  • DescriptionSodium Formate (Cryst./Certified ACS) (500g)
Storage Code Gray

Key Features

  • Assay (HCOONa) - ≥99.0%
  • Insoluble Matter - ≤0.005%
  • Chloride - ≤0.001%
  • Sulfate - ≤0.001%
  • Calcium - ≤0.005%
  • Heavy Metals (as Pb) - ≤5ppm
  • Iron - ≤5ppm

More About this Item


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat