Fisher Chemical SB8221 Barium Chloride Solution (20% w/v) (1 liter)

Representative image only.
Ricca Chemical Company Mfr # SB8221 – Item # EW-88002-72
Quality and consistency your lab can count on
  • Reagents and solutions to meet your lab and research needs
  • Assay (BaCl2·2H2O) — 19.5-20.5% w/v
  • Color (APHA) — ≤10

SPECS & DESCRIPTION

Specifications & Description

  • ClassReagent Salts
  • ChemistrySolutions
  • FormLiquid
  • Volume (mL)1000
  • PackagingGlass bottle
  • DOT InformationClass 6.1, Packing Group III
  • Storage SymbolGray
  • Hazard SymbolHarmful
  • CAS NumberIncludes 10326279 and 7732185
  • UN NumberUN3287
  • DescriptionBarium Chloride Solution (20% w/v) (1 liter)
HarmfulStorage Code Gray

Key Features

  • Reagents and solutions to meet your lab and research needs
  • Assay (BaCl2·2H2O) — 19.5-20.5% w/v
  • Color (APHA) — ≤10

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat