Fisher Chemical Sodium Chromate Anhydrous, Certified Grade, 250 g

Representative image only.
Acros Organics Mfr # S272250 – Item # EW-88023-60
Quality and consistency your lab can count on
  • Reagents and solutions to meet your lab and research needs
  • Sulfate — ≤0.01%
  • Loss on Drying at 105°C — ≤1.0%
  • Chloride — ≤0.010%
  • Aluminum — To pass test
  • Calcium — ≤0.005%

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeCertified
  • Empirical FormulaNa2CrO4
  • ClassReagent Salts
  • ChemistryAnalytical Reagents
  • SynonymsChromic Acid Disodium Salt
  • FormSolid
  • Weight (g)250
  • Formula Weight161.97
  • PackagingGlass Bottle
  • DOT InformationSodium Chromate; Class 6.1 (8), Packing Group II
  • Storage SymbolYellow
  • Hazard SymbolOxidizing, Toxic
  • UN NumberUN3290
  • CAS Number7775-11-3
  • Melting Point C (° C)792
  • DescriptionSodium Chromate Anhydrous, Certified Grade, 250 g
OxidizingToxic

Key Features

  • Reagents and solutions to meet your lab and research needs
  • Sulfate — ≤0.01%
  • Loss on Drying at 105°C — ≤1.0%
  • Chloride — ≤0.010%
  • Aluminum — To pass test
  • Calcium — ≤0.005%

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat