Fisher Chemical Sodium Sulfite Anhydrous, Certified ACS Grade, 500 g

Representative image only.
Fisher Chemical Mfr # S430500 – Item # EW-88026-00
Quality and consistency your lab can count on
  • Reagents and solutions to meet your lab and research needs
  • Assay — ≥98.0%
  • Insoluble Matter — ≤0.005%
  • Free Acid — To pass test
  • Titratable Free Base — ≤0.03mEq/g
  • Chloride — ≤0.02%
  • Heavy Metals (as Pb) — ≤0.001%
  • Iron — ≤0.001%

$122.50USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeCertified ACS
  • Empirical FormulaNa2SO3
  • ClassReagent Salts
  • ChemistryAnalytical Reagents
  • FormSolid
  • SynonymsDisodium sulfite, Sulfuqrous acid, disodium salt
  • Weight (g)500
  • Formula Weight126.04
  • PackagingGlass Bottle
  • DOT InformationNot regulated by DOT
  • Storage SymbolGrey
  • CAS Number7757-83-7
  • Melting Point C (° C)500
  • DescriptionSodium Sulfite Anhydrous, Certified ACS Grade, 500 g
HarmfulStorage Code Gray

Key Features

  • Reagents and solutions to meet your lab and research needs
  • Assay — ≥98.0%
  • Insoluble Matter — ≤0.005%
  • Free Acid — To pass test
  • Titratable Free Base — ≤0.03mEq/g
  • Chloride — ≤0.02%
  • Heavy Metals (as Pb) — ≤0.001%
  • Iron — ≤0.001%

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat