Fisher Chemical SS267 Sodium Hydroxide 10N Concentrate (Certified) (100 mL)

Representative image only.
Fisher Chemical Mfr # SS267 – Item # EW-88085-27
Quality and consistency your lab can count on
  • Reagents and solutions to meet your lab and research needs
  • Normality when Diluted to 1 Liter - 0.990-1.010
  • Color (APHA) - ≤10

SPECS & DESCRIPTION

Specifications & Description

  • Normality10
  • Purity GradeCertified
  • ClassReagent Acids and Bases
  • ChemistrySolutions
  • FormLiquid
  • SynonymsCaustic Soda Solution
  • Volume (mL)100
  • PackagingPoly Bottle
  • DOT InformationSodium Hydroxide Solution; Class 8, Packing Group II
  • Storage SymbolWhite
  • Hazard SymbolCorrosive
  • CAS Number1310-73-2 and 7732-18-5
  • UN NumberUN1824
  • DescriptionSodium Hydroxide 10N Concentrate (Certified) (100 mL)
CorrosiveStorage Code White

Key Features

  • Reagents and solutions to meet your lab and research needs
  • Normality when Diluted to 1 Liter - 0.990-1.010
  • Color (APHA) - ≤10

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat