Fisher Chemical SS73 Silver Nitrate Concentrate (Certified) (100mL)

Representative image only.
Fisher Chemical Mfr # SS73 – Item # EW-88085-50
Quality and consistency your lab can count on
  • Reagents and solutions to meet your lab and research needs

SPECS & DESCRIPTION

Specifications & Description

  • Normality1
  • Purity GradeCertified
  • Empirical FormulaAgNO3
  • Linear FormulaAgNO3
  • ClassGeneral Solutions
  • ChemistrySolutions
  • FormLiquid
  • SynonymsNitric acid; silver salt solution
  • Volume (mL)100
  • Formula Weight169.87
  • PackagingAmber Glass
  • DOT InformationENVIRONMENTALLY HAZARDOUS SUBSTANCES, LIQUID, N.O.S.; Class 9, Packing Group III
  • Storage SymbolYellow
  • Hazard SymbolCorrosive, Environment
  • CAS Number7761-88-8 and 7732-18-5
  • UN NumberUN3082
  • DescriptionSilver Nitrate Concentrate (Certified) (100 mL)
Corrosive

Key Features

  • Reagents and solutions to meet your lab and research needs

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat