Fisher Chemical SX164 ScintiVerse E Cocktail (Scintanalyzed) (4 liter)

Representative image only.
Fisher Chemical Mfr # SX164 – Item # EW-88085-72
  • Specific Gravity at 25°C - 0.930-0.936
  • Water - ≤0.7%
  • LSC Analysis:
  • &tab;Background - ≤50cpm
  • &tab;3H Efficiency - ≥0.65
  • &tab;Quenched Efficiency Ratio, 0.5mL H2O - ≥0.55
  • &tab;Quenched Efficiency Ratio, 1.0mL H2O - ≥0.50
  • &tab;Quenched Efficiency Ratio, 2.0mL H2O - ≥0.45
  • &tab;Quenched Efficiency Ratio, 3.0mL H2O - ≥0.40

SPECS & DESCRIPTION

Specifications & Description

  • ChemistrySolutions
  • Volume (mL)4000
  • CAS Number1330-20-7
  • DescriptionScintiVerse E Cocktail (Scintanalyzed) (4 liter)
HarmfulStorage Code Red

Key Features

  • Specific Gravity at 25°C - 0.930-0.936
  • Water - ≤0.7%
  • LSC Analysis:
  • &tab;Background - ≤50cpm
  • &tab;3H Efficiency - ≥0.65
  • &tab;Quenched Efficiency Ratio, 0.5mL H2O - ≥0.55
  • &tab;Quenched Efficiency Ratio, 1.0mL H2O - ≥0.50
  • &tab;Quenched Efficiency Ratio, 2.0mL H2O - ≥0.45
  • &tab;Quenched Efficiency Ratio, 3.0mL H2O - ≥0.40

More About this Item


Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat