Fisher Chemical SX18250 ScintiVerse BD Cocktail (Scintanalyzed) (250 mL)

Representative image only.
Fisher Chemical Mfr # SX18250 – Item # EW-88085-74

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeScintanalyzed
  • ClassGeneral Solutions
  • ChemistrySolutions
  • FormLiquid
  • Volume (mL)250
  • PackagingGlass bottle
  • DOT InformationNot regulated by DOT
  • Storage SymbolGray
  • CAS Number67774-74-7, 577-11-7, 9016-45-9, 92-71-7, 13280-61-0 and 3806-34-6
  • DescriptionScintiVerse BD Cocktail (Scintanalyzed) (250 mL)

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat