Fisher Chemical Tetrahydrofuran, Histological Grade, 500 mL

Representative image only.
Fisher Chemical Mfr # T400500 – Item # EW-88040-24
Quality and consistency your lab can count on
  • Reagents and solutions to meet your lab and research needs
  • Water — ≤1%
  • Residue after Evaporation — To pass test
  • Dilution Test — To pass test

PRODUCT OPTIONS

$140.50USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeHistological
  • Empirical FormulaCH2(CH2)2CH2O
  • ClassReagent Solvents
  • ChemistrySolvents
  • FormLiquid
  • SynonymsTHF
  • Volume (mL)500
  • Formula Weight72.11
  • PackagingGlass Bottle
  • DOT InformationTetrahydrofuran; Class 3, Packing Group II
  • Storage SymbolRed
  • Hazard SymbolHighly Flammable, Irritant
  • UN NumberUN2056
  • CAS Number109-99-9
  • Boiling Point C (° C)66
  • Melting Point C (° C)-108.4
  • DescriptionTetrahydrofuran, Histological Grade, 500 mL
FlammableIrritantStorage Code Red

Key Features

  • Reagents and solutions to meet your lab and research needs
  • Water — ≤1%
  • Residue after Evaporation — To pass test
  • Dilution Test — To pass test

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat