Fisher Chemical Water, Deionized Ultra-Filtered, 20 L

Representative image only.
Fisher Chemical Mfr # W220 – Item # EW-88042-62
Quality and consistency your lab can count on
  • Water meets DIUF standards
  • Water comes in 20 L size and packaged in a PolyPac™

PRODUCT OPTIONS

$351.50USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradeDeionized Ultra-Filtered (DIUF)
  • Empirical FormulaH2O
  • Synonymsdihydrogen oxide, dihydrogen monoxide
  • FormLiquid
  • Volume (mL)20000
  • Formula Weight18.02
  • PackagingPolyPac™
  • DOT InformationNot regulated by DOT
  • CAS Number7732-18-5
  • Boiling Point C (° C)100
  • Boiling Point F (° F)212
  • DescriptionWater, Deionized Ultra-Filtered, 20 L
Storage Code Gray

Key Features

  • Water meets DIUF standards
  • Water comes in 20 L size and packaged in a PolyPac™

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat