Fisher Cleaning Solution, Chromic-Sulfuric Acid, 500 mL

Representative image only.
Fisher Mfr # SC88500 – Item # EW-88005-83
Removes nearly impossible stains from glassware
  • Quality and consistency your lab can count on

$123.00USD / Each
Often bought with…

SPECS & DESCRIPTION

Specifications & Description

  • ConcentrationChromium Trioxide 0.5%, Sulfuric Acid 93.87%, Water 5.63%
  • ClassCleaning Solution
  • ChemistryAcids
  • FormLiquid
  • Volume (mL)500
  • PackagingSafe-Cote™ plastic bottle
  • CAS Number1333-82-0, 7664-93-9, and 7732-18-5
  • Boiling Point C (° C)290
  • DescriptionCleaning Solution, Chromic-Sulfuric Acid, 500 mL
CorrosiveStorage Code White

Key Features

  • Quality and consistency your lab can count on

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat