Fisher Research Labs XLT-17 Leak Detector Kit For Gas And Liquid
Representative image only.
Fisher Research Labs Mfr # XLT-17 – Item # EW-20001-56
Detects and pinpoints liquid leaks in pipes
  • 4 frequency filters: low pass, high pass, band pass and all pass
  • Last setting memory feature
  • Rugged membrane keypad
  • Shielded sensor for reduced response to outside noise
  • Improved headset for clearer sound response
  • Automatic battery test—real time voltage level; operates on two 9 V batteries
  • Compact control box that can be worn around the neck or over the shoulder with an attachable strap
  • 2 year warranty
$2,602.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionLeak Detector Kit For Gas And Liquid
  • Warranty2 Years
Two year warranty
Key Features
  • 4 frequency filters: low pass, high pass, band pass and all pass
  • Last setting memory feature
  • Rugged membrane keypad
  • Shielded sensor for reduced response to outside noise
  • Improved headset for clearer sound response
  • Automatic battery test—real time voltage level; operates on two 9 V batteries
  • Compact control box that can be worn around the neck or over the shoulder with an attachable strap
  • 2 year warranty
More About this Item
This liquid leak detector finds leaks in four easy steps: by electronically amplifying leak sounds, selectively filtering out noise, isolating the leak sound, and leading the operator to the source of the leak.