Fitting Manifold, Polycarbonate, Four Ports, Female Luer Locks, 180° Rotation
Representative image only.
Item # EW-06464-85
Regulate flow without any leaking
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
PRODUCT OPTIONS
$16.10USD / Each
BUY MORE & SAVE
  • 1-9$16.10USD / Each
  • 10-49$14.49USD / Each
  • 50-99$13.69USD / Each
  • 100+$12.88USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Outlet PortFemale Luer
  • Inlet PortFemale Luer
  • Wetted MaterialsPolycarbonate
  • Flow Pattern4 Ports, 180 Degree Rotation
  • DescriptionPolycarbonate individual manifolds with luer locks; 4 ports; 180° rotation
Key Features
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
More About this Item
Polycarbonate manifolds have polyurethane stems and HDPE handles. Ports have female luer locks.