Fitting, Threaded Nipple NPT(M), PFA, 3/4" male
Representative image only.
Item # EW-06546-31
Maintain a permanent connection
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
PRODUCT OPTIONS
$37.00USD / Each
BUY MORE & SAVE
  • 1-9$37.00USD / Each
  • 10-49$33.30USD / Each
  • 50-99$31.45USD / Each
  • 100+$29.60USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialPFA
  • Sterilization MethodAutoclave or ethylene oxide (ETO) only
  • Connection TypeNPT(M)
  • Thread size NPT(M) (port 1) (in)3/4
  • Thread size NPT(M) (port 2) (in)3/4
  • DescriptionFitting, Threaded Nipple NPT(M), PFA, 3/4" male
Key Features
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
More About this Item
Threaded fittings provide a more permanent fluid path. Choose the size and material to meet your application needs.