Flex Spout Adapter for 5-Gallon Containers with 2-5/8" Opening, polyethylene
Representative image only.
Item # EW-07079-15
Compatible with Siphon and Piston Hand-Operated Pumps
  • Choose from a wide range of replacement parts and accessories
PRODUCT OPTIONS
$20.50USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionFlex Spout Adapter for 5-Gallon Containers with 2-5/8" Opening, polyethylene
Key Features
  • Choose from a wide range of replacement parts and accessories
More About this Item
Model 07079-10 steel screw in adapter features a 1-3/4" NPT(F) connection and fits 5 gallon pails.
Model 07079-15 flex spout adapter is constructed of durable polyethylene and fits 5 gallon containers with 5-5/8" (6.7 cm) openings.
Model 07079-17 bung converter kit converts a Mauser course bung thread or standard course buttress to a 2" NPT connection.