Flir 55001-0102 Automation Thermal Camera (640x480), F=24.6mm and 25 Deg Lens
Representative image only.
Flir Mfr # FLIR A615 W/25 LENS – Item # EW-39755-76
  • 640 x 480 pixel IR resolution
  • MPEG-4 streamed video
  • C-Video
  • Extensive analysis functions
  • Built in alarm functions
  • Ethernet/IP connectivity
  • Digital inputs/outputs
  • 16-bit temperature linear output
  • GigE Vision™ standard compatibility
  • GenICam™ protocol support
  • Windowing mode
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Min Temperature (° F)-4
  • Max Temperature (° F)3632
  • Min Temperature (° C)-20
  • Max Temperature (° C)2000
  • Thermal Sensitivity (N.E.T.D) (° F)≤ 0.09 at 86
  • Thermal Sensitivity (N.E.T.D) (° C)≤ 0.05 at 30
  • Camera Included A615 (640x480), f=24.6mm and 25 deg Lens
  • DescriptionAutomation Thermal Camera (640x480), F=24.6mm and 25 Deg Lens
  • Warranty1 year
1 Year WarrantyConformite Europeenne
Key Features
  • 640 x 480 pixel IR resolution
  • MPEG-4 streamed video
  • C-Video
  • Extensive analysis functions
  • Built in alarm functions
  • Ethernet/IP connectivity
  • Digital inputs/outputs
  • 16-bit temperature linear output
  • GigE Vision™ standard compatibility
  • GenICam™ protocol support
  • Windowing mode
  • 1 year warranty
More About this Item
FLIR A615 targets R&D and automation applications and is a perfect instrument for industries which rely on permanent monitoring of objects that are fast moving or when the temperature changes over time is quiet fast. FLIR A615 also complies with standards like GigE Vision and GenICam. Due to its compliance to standards, FLIR A615 is a Plug&Play device within 3rd parties Machine Vision softwares like NIs IMAQ Vision TM and the MVTecs Halcon TM software. Examples of applications are PCB testing, validation and design verification within the automotive, medical and power electronics industries.
Package Contents:
* Cardboard box
* Infrared camera with lens
* Calibration certificate
* Ethernet cable
* Mains cable
* Power cable, pig-tailed
* Power supply
* Printed Getting Started Guide
* Printed Important Information Guide
* USB cable
* User documentation CD-ROM
* Utility CD-ROM
* Warranty extension card or Registration card
Available for sale in the United States and Canada only