Flir 61201-1004 Fixed Automation Thermal Camera IP-66 Rated with 45 Deg Lens
Representative image only.
Flir Mfr # FLIR A300F W/45 LENS – Item # EW-39755-34
Accurate temperature readings at an affordable price
  • Housing meets IP-66 requirements
  • Produces 320 x 240 pixel IR resolution thermal images
  • MPEG-4 streamed video
  • C-video
  • 45 degree lens
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Thermal Sensitivity (N.E.T.D) (° F)≤ 0.09 at 86
  • Thermal Sensitivity (N.E.T.D) (° C)≤ 0.05 at 30
  • Camera Included A300f 45 Fixed IP-66 Rated with 45 deg Lens
  • DescriptionFixed Automation Thermal Camera IP-66 Rated with 45 Deg Lens
  • Warranty1 year
1 Year WarrantyConformite EuropeenneIP 66 Rated
Key Features
  • Housing meets IP-66 requirements
  • Produces 320 x 240 pixel IR resolution thermal images
  • MPEG-4 streamed video
  • C-video
  • 45 degree lens
  • 1 year warranty
More About this Item
The FLIR fix mount thermal cameras offer an affordable and accurate temperature measurement solution for anyone who needs to solve problems that do not call for the highest speed or reaction, and who uses a PC. Due to its composite video output, it is an excellent choice for thermal image automation applications, where you can utilize its unique properties such as looking through steam.
Package contents include:
• Infrared camera with lens
• Calibration certificate
• Cardboard box
• Downloads brochure
• Ethernet cable
• FLIR apps card
• FLIR Tools download card
• Getting Started Guide
• Important Information Guide
• Mains cable
• Optics brochure
• Power cable, pig-tailed
• Power supply
• Service and training brochure
• Thank you card
• User documentation CD-ROM
• Utility CD-ROM
• Registration card
Automation thermal imagers in many cases require application support prior to ordering. Please contact our technical application group or sales team for support on this product. In some areas of the country onsite field assessments can be arranged.
Available for sale in the United States and Canada only