Flir 62103-RPP Restoration Professional Package with T420 Thermal Imager
Representative image only.
Extech Mfr # 62103-RPP – Item # EW-39756-31
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Pixels / Thermal Resolution76,800
  • Detector Size320 x 240
  • Min Temperature (° F)-4
  • Max Temperature (° F)1202
  • Min Temperature (° C)-20
  • Max Temperature (° C)650
  • ZoomYes
  • Thermal Sensitivity (N.E.T.D) (° F)≤ 0.08 at 86
  • Thermal Sensitivity (N.E.T.D) (° C)≤ 0.045 at 30
  • Focus TypeManual, Automatic
  • Image ModesYes
  • Fusion (Picture in Picture)Yes
  • Enhanced Spectral ImagingYes
  • Software IncludedYes
  • Display TypeLCD
  • Low Battery IndicatorYes
  • Battery life4 hours
  • Height (in)16
  • Width (in)24
  • Length (in)16
  • DescriptionRestoration Professional Package with T420 Thermal Imager
  • Warranty2 year
Two year warrantyConformite Europeenne
Key Features
  • 2 year warranty
More About this Item
FLIR 62103-RPP Restoration Pro Package; Includes: 62103-1101 Model T420 High Performance Thermal Imaging Infrared Camera, MR01 Replacement Temp/RH Sensor, 1910490 Car Charger, 1196398 Extra Battery, T198495 Pouch Case, T911093 Tool Belt, T198583 FLIR Tools+ Scratchcard and VP50 Non-Contact Voltage (NCV) Detector + Flashlight; Temperature range from -4°F to 482°F (-20°C to 250°C); Sharp thermal resolution at 76800 pixels for solid accuracy from farther away; Integrated IR Thermometer provides fast non-contact surface temperature measurements; Safe CAT IV-1000V rating and toolbox tough with rubber reinforced case and buttons, and a low-profile detection tip; Vibration and red LED alarms alert users to the presence of voltage in noisy areas; Versatile High/Low sensitivity modes detect voltage on industrial equipment and low-voltage installations