Flir CM174 Clamp Meter with IGM, 600 A AC/DC

Representative image only.
Flir Mfr # CM174 – Item # EW-20050-92
Visually identify electrical issues with the first thermal imaging clamp meter!
  • Infrared Guided Measurement (IGM™) technology allows you to find issues faster
  • Easily confirm findings with accurate amperage and voltage measurements, and center-point temperature readings
  • Compact form-factor and narrow jaw permit easy access to difficult locations
  • Work safely—scan a panel or cabinet for hazards using IGM without direct contact
  • Verify problems, check load, and validate hot spots quickly and easily
  • Center-point temperature confirms area hot spots
  • Laser and crosshair pinpoint problem locations through thermal images
  • Limited Lifetime Warranty

SPECS & DESCRIPTION

Specifications & Description

  • DC Min (Volts)0.1
  • DC Max (Volts)1000
  • DC Min (amps)0.1
  • DC Max (amps)600
  • Current (AC/DC)0.1 to 600 A
  • AC Min (Volts)0.1
  • AC Max (Volts)1000
  • AC Min (amps)0.1
  • AC Max (amps)600
  • AC Current Accuracy±2.0%
  • Resistance Min (Ω)0.1
  • Resistance Max (Ω)6000
  • Capacitance Min (mF)0
  • Capacitance Max (mF)1000
  • Frequency Min (KHz)0
  • Frequency Max (KHz)60
  • Min Temperature (° F)-13
  • Max Temperature (° F)302
  • Min Temperature (° C)-25
  • Max Temperature (° C)150
  • Temperature Accuracy±5.4⁰F (3⁰C) or ±3% of reading
  • Display6000 Count 2.0 in (50 mm) Color TFT
  • Jaw Size1.38
  • PowerFour AAA batteries
  • Width (in)4.3125
  • Length (in)10.625
  • Height (in)2
  • DescriptionClamp Meter with IGM, 600 A AC/DC
  • WarrantyLTD Life Time
Limited Lifetime WarrantyUL Listed.

Key Features

  • Infrared Guided Measurement (IGM™) technology allows you to find issues faster
  • Easily confirm findings with accurate amperage and voltage measurements, and center-point temperature readings
  • Compact form-factor and narrow jaw permit easy access to difficult locations
  • Work safely—scan a panel or cabinet for hazards using IGM without direct contact
  • Verify problems, check load, and validate hot spots quickly and easily
  • Center-point temperature confirms area hot spots
  • Laser and crosshair pinpoint problem locations through thermal images
  • Limited Lifetime Warranty

More About this Item

The FLIR CM174 imaging clamp meter with IGM is the first clamp meter equipped with a built-in thermal imager, allowing you to quickly identify problems you can’t see with a standard clamp meter. Infrared Guided Measurement (IGM) technology visually guides you to the precise location of a potential electrical problem, identifying dangerous and unknown problem areas safely.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat