Flir CM57-2 Flexible 18-inch Clamp Meter
Representative image only.
Flir Mfr # CM57-2 – Item # EW-39758-55
With its 18-inch (45.72 cm) flexible coil clamp, the CM57-2 allows you to take measurements in tight spots, making it ideal for multiple-conductor measurements and double-wrap requirements
  • Wrap the flexible coil around multiple conductors or large cables easily
  • See clearly in any condition with dual LED worklights that provide better visibility in dark or poorly lit areas
  • Stream live data in real time to your mobile device using the METERLiNK® app
$288.99USD / Each
$339.99USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • AC Max (amps)3000
  • AC Current Accuracy±3.0% + 5 digits
  • Display3000-count LCD with backlight and multi‐function indicators
  • Memory20,000 points, 1 minute sample rate
  • InterfaceBluetooth
  • Bandwidth45 Hz to 500 Hz (sine wave)
  • Safety RatingCAT IV-600 V, CAT III-1000 V
  • Power2 × 1.5 V AAA
  • DescriptionFlexible 18-inch Clamp Meter
Key Features
  • Wrap the flexible coil around multiple conductors or large cables easily
  • See clearly in any condition with dual LED worklights that provide better visibility in dark or poorly lit areas
  • Stream live data in real time to your mobile device using the METERLiNK® app
More About this Item
The FLIR CM57-2 is a flexible clamp meter designed for difficult current measurements. The meter also features dual LED work lights for better visibility and AC current ranges from 30.00 A to 3000 A. Stream live data in real time from the CM57-2 to your mobile device over Bluetooth using the METERLiNK® app to visually compare log files, customize alarm thresholds, create reports, and share readings instantly with team members.