Flir Cx5 Hazardous Location-Rated Thermal Camera

Representative image only.
Flir Mfr # 89401-0203 – Item # EW-39758-52
Safely inspect heat sources and identify issues in explosive environments
  • Eliminates the need for hot work permits in hazardous locations due to gas, vapor, and dust
  • MSX® imagery enhancement provides sharper, easier to analyze images
  • Measures up to 400 °C (752 °F)
  • Detects temperature differences down to 70 mK
  • Lightweight and portable

$1,999.00USD / Each
sign in for your price.

SPECS & DESCRIPTION

Specifications & Description

  • Pixels / Thermal Resolution160 × 120 (19,200 pixels)
  • Detector Speed (Hz)8.7
  • Field Of View (FOV)54° × 42°
  • Min Temperature (° F)-4
  • Max Temperature (° F)752
  • Min Temperature (° C)-20
  • Max Temperature (° C)400
  • AccuracyAt ambient temp. 15°C to 35°C (59°F to 95°F) and object temp. above 0°C (32°F), 0°C to 100°C (32°F to 212°F): ±5°C (±9°F), 100°C to 400°C (212°F to 752°F): ±5%
  • Focus TypeFocus free
  • Palette SelectionIron, gray, rainbow, arctic, lava, rainbow HC
  • Image ModesInfrared image, visual image, MSX, and Picture-in-picture (IR area on visual image)
  • Memory Storage TypeInternal memory and onboard FLIR Ignite cloud connectivity (with wi-fi)
  • Memory Capacity>5000 images
  • Display TypeCapacitive touchscreen
  • Battery life4 hours
  • BatteryRechargeable built-in Lithium ion battery
  • Height (in)3 3/4
  • Width (in)3 7/8
  • Length (in)9 1/4
  • DescriptionFLIR Cx5 Hazardous Location-Rated Thermal Camera

Key Features

  • Eliminates the need for hot work permits in hazardous locations due to gas, vapor, and dust
  • MSX® imagery enhancement provides sharper, easier to analyze images
  • Measures up to 400 °C (752 °F)
  • Detects temperature differences down to 70 mK
  • Lightweight and portable
  • Instantly upload images to the cloud
  • Battery lasts up to 4 hours

More About this Item

The Flir Cx5 thermal imaging camera is built to provide safe inspection in hazardous locations such as chemical, oil and gas, and wastewater treatment facilities where combustible gases are present. The Cx5 camera is certified IEC Ex IIC T4 II Gc II 3 G and Ex tc IIB T135°C Dc, eliminating the need for hot work permits while on site. Its tough enclosure prevents any potential ignition inside from entering the atmosphere.

Accurately measure temperatures of nearby targets between -20 to 400 °C (-4 to 752 °F). The 160 × 120 (19,200 pixels) thermal resolution camera, 3.5" touchscreen interface, and MSX® (Multi-Spectral Dynamic Imaging) feature allow you to see the temperature gradients instantly. The MSX imagery enhancement feature adds key details from the onboard visible light camera to create a sharper, more enhanced image.

This lightweight, pocket-sized camera only weighs just over 1.5 lb and measures 23.5 x 10 x 9.5 cm so you can easily carry it around and discover hot spots and signs of degrading insulation. The camera housing conforms to IP54 (IEC 60529) standard and can safely withstand a drop of 2 m (6.6 ft). Upload images from your Cx5 to the cloud using FLIR Ignite™ cloud service, where you can easily edit, organize, store, and share data.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat