DescriptionCompact Thermal Imaging Camera with Extended Temperature Range, MSX, and WiFi; 19,200 Pixels
Includes
Hard carry case, USB mini-B cable, AC adapter/charger with EU, UK, US, and AU plugs, and 1 rechargeable lithium-ion battery.
Key Features
Connect over Wi-Fi to mobile devices
USB interface for easy data transmission
Get the most out of your data with user-friendly mobile app
Exclusive MSX thermal imaging technology gives more detailed images
Minimize fatigue with easy-grip handle
Get hours of operation with robust, rechargeable Li-ion battery
Meets IP54 dust/splashproof standards
Resolution up to 160 x 120 infrared pixels
2 year warranty
More About this Item
The FLIR® Ex-series point-and-shoot thermal imagers accurately measure temperatures using the center spot or area box with max/min. With the highest accuracy for this class of thermal imagers at ±2%, these imagers are perfect for identifying hot spots or a rise in temperature which often indicates a problem exists, or a failure could be imminent. Ideal for use in applications where a visual indication of temperature variation may be helpful, such as troubleshooting electrical systems and machinery, or conducting a home energy audit.
All E-Series thermal imagers feature Wi-Fi connectivity to your Apple® or Android™ mobile device. Simply download the FLIR Tool Mobile app and link it to your thermal imager. Images can be transferred to your mobile device via Wi-Fi or USB cable for further analysis, editing, creating reports, and e-mailing results.
The Multi-Spectral Dynamic Imaging System (MSX) feature on the thermal imagers provide extraordinary real-time thermal detail for better perspective and interpretation of problem areas. It eliminates the need to refer back to a visual image. Other features include a large LCD, selectable on-screen viewing modes, focus-free lens for convenient close-focus viewing (from 1.6 feet to infinity), automatic and manual temperature scale modes, internal memory that stores up to 500 images with embedded temperature data, and five image modes: thermal MSX, thermal, picture-in-picture, thermal blending, and digital camera.
Extremely lightweight and compact design with easy-grip handle results in less user fatigue. Rugged construction meets IP54 dust/splashproof standards and withstands a drop from 2 meters. Designed for continuous operation on a single charge, the unit features a Li-ion rechargeable battery with AC adapter/charger, which produces long battery life.
Available for sale in the United States and Canada only.