Flir IRW-SS-2C Infrared Inspection Window, Stainless Steel, 2"

Representative image only.
Flir Mfr # IRW-SS-2C – Item # EW-39756-32
Helps reduce the vast majority of dangerous arc flash occurrences
  • Rugged frame handles both outdoor and indoor use
  • Decrease downtime and increase efficiency with easy installation
  • Easy flip-open hatch for faster inspections
  • Simple thumb screw releases the permanently hinged cover
  • Beaker features a 50 mL capacity
  • Designed with graduation range from 10 to 40 mL in 10 mL intervals
  • Features yellow colored markings
  • Limited Lifetime Warranty

SPECS & DESCRIPTION

Specifications & Description

  • Max Temperature (° F)500
  • Max Temperature (° C)260
  • Camera IncludedNo
  • Height (in)3 23/64
  • Width (in)2 7/8
  • Length (in)1
  • DescriptionInfrared Inspection Window, Stainless Steel, 2\"
  • WarrantyLimited Lifetime
IP 67Limited Lifetime Warranty

Includes

window with hinged cover, gasket, and ring nut.

Key Features

  • Rugged frame handles both outdoor and indoor use
  • Decrease downtime and increase efficiency with easy installation
  • Easy flip-open hatch for faster inspections
  • Simple thumb screw releases the permanently hinged cover
  • Beaker features a 50 mL capacity
  • Designed with graduation range from 10 to 40 mL in 10 mL intervals
  • Features yellow colored markings
  • Limited Lifetime Warranty

More About this Item

Opening electrical cabinets to perform thermal and visual inspections of live components is dangerous work, exposing you to the risk of a hazardous arc flash incident. The FLIR IRW-series of thermal inspection windows help eliminate that danger. The window allows all brands of thermal imagers to capture both visible light and thermal images along with allowing LED and laser illumination to pass through the glass. A secure, permanently hinged cover opens easily with one hand, which means there’s nothing to remove, drop, or lose. The IRW-Series windows are comprehensively tested for safety and have a limited lifetime warranty.

Available for sale in the United States and Canada only

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat