Flir MIO-FCR-1 I/O module for FC-Series R Thermal Cameras
Representative image only.
Flir Mfr # MIO-FCR-1 – Item # EW-39757-26
Monitor up to seven thermal imaging cameras remotely
  • Easily configure each channel for alarm or analog outputs
  • 4 to 20 mA output provides greater process control options
  • Built-in web server simplifies setup
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Length (\")5 3/4
  • Width (InchShort)3 5/8
  • Number of Channels2
  • Height (in)1 5/8
  • Power (VDC)24
  • DescriptionI/O module for FC-Series R Thermal Cameras
  • Warranty1 year
1 Year WarrantyConformite Europeenne
Key Features
  • Easily configure each channel for alarm or analog outputs
  • 4 to 20 mA output provides greater process control options
  • Built-in web server simplifies setup
  • 1 year warranty
More About this Item
Turn your FLIR AX8, A310, and FC-Series R thermal imaging cameras into a complete remote condition/process monitoring and fire detection system with the compact MIO series intelligent I/O modules. Easy to set up, the I/O modules are simple to configure via a built-in web server—no need for a separate PC or custom computer software.
Up to seven cameras can be connected to the I/O modules. Systems can be expanded by adding more I/O modules to the network. Once connected, the module can target designated regions of interest and start monitoring. The module can be programmed through the web server for real-time audible and visual alarms when temperature thresholds are breached. The MIO series can also output an analog 4 to 20 mA current loop signal per each region of interest, providing you with various process control options.
Available for sale in the United States and Canada only.