Flir ONE® Edge Pro Thermal Camera with Wireless Connectivity

Representative image only.
Flir Mfr # 11002-0201 – Item # EW-39758-50
Capture vivid thermal images wirelessly on your iOS® and Android smart devices
  • Remotely inspect hazardous areas up to 30 m (100 ft) away
  • MSX® imagery enhancement provides sharper, easier to analyze images
  • Measures up to 400 °C (752 °F)
  • Detects temperature differences down to 70 mK
  • Instantly upload images or videos to the cloud

$549.99USD / Each
sign in for your price.

SPECS & DESCRIPTION

Specifications & Description

  • Pixels / Thermal Resolution160 × 120 (19,200 pixels)
  • Detector Speed (Hz)8.7
  • Field Of View (FOV)HFOV: 54° ±1°; VFOV: 42° ±1°
  • Min Temperature (° F)-4
  • Max Temperature (° F)752
  • Min Temperature (° C)-20
  • Max Temperature (° C)400
  • Accuracy±3°C (5.4°F) or ±5%. Applicable 60 sec after start-up when the unit is within 15°C to 35°C (59°F to 95°F) and the scene is within 5°C to 120°C (41°F to 248°F)
  • Focus TypeFixed 30 cm to infinity
  • Palette SelectionGray (white hot), hottest, coldest, iron, rainbow, contrast, arctic, lava, and wheel
  • Image ModesThermal, visual, and MSX
  • Visible Light Camera Resolution (Pixels)640 x 480
  • EmissivityMatte, semi-matte, semi-glossy, glossy
  • Video OutputYes
  • Battery life2.5 hours
  • Height (in)1 1/2
  • Width (in)1 3/8
  • Depth (in)5.875
  • DescriptionONE® Edge Pro Thermal Camera with Wireless Connectivity

Key Features

  • Remotely inspect hazardous areas up to 30 m (100 ft) away
  • MSX® imagery enhancement provides sharper, easier to analyze images
  • Measures up to 400 °C (752 °F)
  • Detects temperature differences down to 70 mK
  • Instantly upload images or videos to the cloud
  • Battery lasts up to 2.5 hours

More About this Item

Connect to your smart device wirelessly with The Flir One Edge Pro to capture thermal images. The camera takes high-quality thermal images to help you observe invisible heat sources and identify problem areas. Conveniently scan out-of-reach targets and view the images on your device or clip it to your phone or tablet for one-handed operation. Lightweight and slimline design with a natural grip lets you take images from any position or angle.

Measure temperatures between -20 to 400 °C (-4 to 752 °F) with three on-screen spot thermometers. The 160 x 120 (19,2000 pixels) thermal resolution camera, VividIR™ thermal image processor, and unique Flir MSX (Multi-Spectral Dynamic Imaging) feature allow you to see more details, measure smaller components from far away, and safely work near energized equipment. The MSX imagery enhancement feature adds key details from the onboard visible light camera to create a sharper, more enhanced image. The camera offers nine dynamic color palettes, automatic and manual shutter, and a fixed 30 cm to infinity focus.

Upload images and videos from your ONE Edge Pro to the cloud using FLIR Ignite™ cloud service, where you can easily edit, organize, store, and share data. Battery lasts up to 2.5 hours and can fast charge in 15 minutes to give 40-minute operating time. This camera is IP54 rated and has been drop-tested from 2 meters, making it ideal for industrial environments.

ACCESSORIES

Accessories

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat