Flir T198126 2-Bay Battery Charger

Representative image only.
Flir Mfr # T198126 – Item # EW-39756-97
Enhance your thermal imager’s capabilities
  • Charge a spare set of batteries—2 bays for convenient, simultaneous, charging
  • 1 year warranty

PRODUCT OPTIONS

$253.98USD / Each
$253.99USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Length (")3 1/5
  • Width (in)6 1/2;6 1/2
  • Height (in)9 19/32;9 19/32
  • Length (in)3 1/5
  • Depth (in)6.5
  • Description2-Bay Battery Charger
  • Warranty1 year
1 Year Warranty

Key Features

  • Charge a spare set of batteries—2 bays for convenient, simultaneous, charging
  • 1 year warranty

More About this Item

A wide offering of Flir accessories to enhance your temperature measurement experience, and meet your application needs.

Available for sale in the United States and Canada only

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat