Flir T911631ACC USB 2.0 A to USB Type-C Cable, 0.9 m for use with Exx / T5xx Imagers

Representative image only.
Flir Mfr # T911631ACC – Item # EW-39757-55
Enhance your thermal imager’s capabilities

PRODUCT OPTIONS

$33.98USD / Each
$33.99USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Height (in)2 45/64
  • Width (in)2 45/64
  • Length (in)2 45/64
  • Depth (in)2.3
  • DescriptionUSB 2.0 A to USB Type-C Cable, 0.9 m for use with Exx / T5xx Imagers

More About this Item

A wide offering of Flir accessories to enhance your temperature measurement experience, and meet your application needs.

Available for sale in the United States and Canada only

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat