Flir TA72 Universal Flexible Current Probe, 3000 A; 2.4" Conductor Diameter
Representative image only.
Flir Mfr # TA72 – Item # EW-20048-92
Simplify current measurements
  • Snake the flexible coil around obstacles — tricky conductors and crowded environments are no longer an issue
  • Connect to your existing clamp meter — the banana plugs are compatible with most DMMs and clamp meters
  • Select your AC current range from 30, 300, or 3000 A
  • See what you''re working on — integrated LEDs illuminate dark cabinets
PRODUCT OPTIONS
$155.55USD / Each
$183.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • AC Min (amps)30
  • AC Max (amps)3000
  • AC Current Accuracy±3.0% + 5
  • Max Clamp Size2.4
  • Safety RatingCAT IV 600V, CAT III 1000V
  • PowerTwo AAA batteries (included)
  • DescriptionUniversal Flexible Current Probe, 3000 A; 2.4" Conductor Diameter
Two year warranty
Includes
Two AAA batteries.
Key Features
  • Snake the flexible coil around obstacles — tricky conductors and crowded environments are no longer an issue
  • Connect to your existing clamp meter — the banana plugs are compatible with most DMMs and clamp meters
  • Select your AC current range from 30, 300, or 3000 A
  • See what you''re working on — integrated LEDs illuminate dark cabinets
More About this Item
The TA72 is a rugged, lightweight current probe, compatible with most digital multimeters and clamp meters accepting standard banana plug connectors. The 10" (25.4 cm) long, flexible-coil jaw can be wrapped around conductors in awkward and tight spaces with ease. With the press of a button the coil is unlocked, letting you conveniently loop the flexible coil around the conductors to be analyzed. Two built-in white, LED lights illuminate workspaces for safe coil placement in dark environments, and the switchable current range is indicated by lights on the clamp handle.