Float Switch with Piggyback Plug; 115 VAC
Representative image only.
Item # EW-07187-40
Convert any manually controlled pump up to 1/2 hp into automatic operation
  • Constructed of PVC and neoprene
$78.20USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Wetted MaterialPVC and neoprene
  • Min Secific Gravity1
  • Max Process Temperature (° C)60
  • Height (in)9.5625
  • Diameter (in)3.0625
  • DescriptionFloat Switch with Piggyback Plug; 115 VAC
Includes
10-ft (3-m) cord with piggyback plug.
Key Features
  • Constructed of PVC and neoprene
More About this Item
Use this float switch to control submersible pumps that don''t have any automatic control. Simply plug the pump''s power cord into the back of the float switch plug (piggyback) and insert the float switch plug into a power outlet. The float switch will control the power to the pump based on the position of the float. Note: Must be used with pumps with a three-prong grounded power plug.