Flowline GT10-5305 Thermal Flow Switch for Gases, Short PVDF Sensor
Representative image only.
Flowline Mfr # GT10-5305 – Item # EW-32756-02
  • Liquid switches are ideal for pump and process protection; gas switches for filter and process protection.
  • Sturdy sensor withstands high flow velocities.
  • Sensor, cable, and junction box are NEMA 4X rated—ideal for corrosive environments.
  • FET switch output interfaces with PLC, SCADA, or alarms.
  • Short sensors can be used in pipe diameters from 3/4" to 1 1/2"; long sensors from 2" to 16".
PRODUCT OPTIONS
$405.00USD / Each
$310.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Repeatability±5% of set point
  • Max Operating Temperature (° F)140
  • Max Operating Temperature (° C)60
  • Display TypeLED
  • DescriptionThermal Flow Switch for Gases, Short PVDF Sensor
Key Features
  • Liquid switches are ideal for pump and process protection; gas switches for filter and process protection.
  • Sturdy sensor withstands high flow velocities.
  • Sensor, cable, and junction box are NEMA 4X rated—ideal for corrosive environments.
  • FET switch output interfaces with PLC, SCADA, or alarms.
  • Short sensors can be used in pipe diameters from 3/4" to 1 1/2"; long sensors from 2" to 16".
  • Easily adjust set point with a single turn of the adjustment knob
  • Switch state is selectable from normally open or normally closed.