Flowline LO10-2305 Optic Leak Detection Pfa Sensor Short 12-36VDC
Representative image only.
Flowline Mfr # LO10-2305 – Item # EW-43312-03
$295.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Power (VDC)12 to 36
  • DescriptionOptic Leak Detection Pfa Sensor Short 12-36VDC
More About this Item
The optic switch is an excellent choice for leak detection applications in and around the secondary containment vessel of sumps, tanks and pipes. The sensor can be applied in a broad range of liquids including sulfuric acid, jet fuel and de-ionized water. The relay output provides a reliable switch interface with remote devices such as a PLC, SCADA or alarm. For maximum flexibility, the sensor is offered in general purpose and intrinsically safe versions with two probe lengths and PP or PFA PTFE materials.